Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC10A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081738.1 | NP_001075207.1 | 916 | 82 | ||
GenBank mRNA | BC149199.1 | - | 4 - 5 | 975 | 82 | |
EF186076.1 | - | 916 | 82 | |||
EF495205.1 | - | 532 | 82 |
Gene Symbol: C6H4orf36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | EF495206.1 | - | 937 | 82 |
Gene Symbol: LOC112441472 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | EF495204.1 | - | 740 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |