Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VAPB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082423.1 | NP_001075892.1 | 2 - 3 | 399 | 141 | |
GenBank mRNA | AB099087.1 | - | 2 - 3 | 331 | 141 | |
BC133497.1 | - | 2 - 3 | 399 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |