Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMTR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082430.2 | NP_001075899.1 | 12 - 13 | 1450 | 63 | |
GenBank mRNA | BC133559.1 | - | 12 - 13 | 1451 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |