Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMTR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082430.2 | NP_001075899.1 | 21 - 22 | 2274 | 68 | |
GenBank mRNA | BC133559.1 | - | 21 - 22 | 2275 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |