Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OCLN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082433.2 | NP_001075902.1 | 6 - 7 | 1485 | 103 | |
GenBank mRNA | AY589500.1 | - | 4 - 5 | 1032 | 103 | |
BC133617.1 | - | 6 - 7 | 1486 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |