Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OCLN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082433.2 | NP_001075902.1 | 8 - 9 | 1702 | 96 | |
GenBank mRNA | BC133617.1 | - | 8 - 9 | 1703 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |