Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIGIRR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082443.2 | NP_001075912.1 | 5 - 6 | 822 | 67 | |
GenBank mRNA | BC133493.1 | - | 5 - 6 | 823 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |