Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSNARE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082446.2 | NP_001075915.1 | 12 - 13 | 1473 | 77 | |
GenBank mRNA | BC133610.1 | - | 12 - 13 | 1475 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |