Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNT11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082456.2 | NP_001075925.1 | 5 - 6 | 1240 | 85 | |
GenBank mRNA | BC133627.1 | - | 5 - 6 | 1241 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |