Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082616.1 | NP_001076085.1 | 8 - 9 | 1334 | 67 | |
GenBank mRNA | BC133393.1 | - | 8 - 9 | 1334 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |