Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD17B7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083375.1 | NP_001076844.1 | 1 - 2 | 48 | 86 | |
GenBank mRNA | BC114716.1 | - | 1 - 2 | 48 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |