Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083383.2 | NP_001076852.1 | 2 - 3 | 402 | 72 | |
GenBank mRNA | BC114872.1 | - | 2 - 3 | 403 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |