Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MICU2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083416.1 | NP_001076885.1 | 11 - 12 | 1199 | 150 | |
GenBank mRNA | BC123699.1 | - | 11 - 12 | 1199 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |