Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXNDC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083420.1 | NP_001076889.1 | 11 - 12 | 2248 | 83 | |
GenBank mRNA | BC123384.1 | - | 11 - 12 | 2248 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |