Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXNDC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083420.1 | NP_001076889.1 | 7 - 8 | 1118 | 65 | |
GenBank mRNA | BC123384.1 | - | 7 - 8 | 1118 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |