Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIF1AN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083443.2 | NP_001076912.1 | 1 - 2 | 192 | 104 | |
GenBank mRNA | BC133607.1 | - | 1 - 2 | 193 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |