Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM98A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083455.1 | NP_001076924.1 | 7 - 8 | 924 | 64 | |
GenBank mRNA | BC123587.1 | - | 7 - 8 | 924 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |