Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPZA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083480.2 | NP_001076949.1 | 3 - 4 | 197 | 88 | |
GenBank mRNA | BC114648.1 | - | 3 - 4 | 201 | 88 | |
BT030727.1 | - | 3 - 4 | 158 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |