Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPZA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083480.2 | NP_001076949.1 | 7 - 8 | 625 | 68 | |
GenBank mRNA | BC114648.1 | - | 7 - 8 | 629 | 68 | |
BT030727.1 | - | 7 - 8 | 586 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |