Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITPKA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083519.1 | NP_001076988.1 | 6 - 7 | 1184 | 86 | |
GenBank mRNA | BC123676.1 | - | 6 - 7 | 1184 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |