Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083639.1 | NP_001077108.1 | 8 - 9 | 1317 | 89 | |
GenBank mRNA | BC134598.1 | - | 8 - 9 | 1317 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |