Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083641.1 | NP_001077110.1 | 5 - 6 | 1353 | 111 | |
GenBank mRNA | BC134626.1 | - | 5 - 6 | 1353 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |