Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDCA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083652.1 | NP_001077121.1 | 5 - 6 | 439 | 63 | |
GenBank mRNA | BC134661.1 | - | 5 - 6 | 439 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |