Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35E3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083654.1 | NP_001077123.1 | 3 - 4 | 854 | 109 | |
GenBank mRNA | BC134617.1 | - | 3 - 4 | 854 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |