Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35E3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083654.1 | NP_001077123.1 | 4 - 5 | 936 | 134 | |
GenBank mRNA | BC134617.1 | - | 4 - 5 | 936 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Inventoried Probe spans exons |