Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTSL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083686.2 | NP_001077155.1 | 2 - 3 | 201 | 60 | |
GenBank mRNA | BC134741.1 | - | 2 - 3 | 203 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |