Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35F6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083704.1 | NP_001077173.1 | 5 - 6 | 721 | 85 | |
GenBank mRNA | BC134546.1 | - | 5 - 6 | 721 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |