Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOC2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083709.1 | NP_001077178.1 | 10 - 11 | 1241 | 124 | |
GenBank mRNA | BC134597.1 | - | 10 - 11 | 1241 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |