Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GEM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083732.2 | NP_001077201.2 | 1 - 2 | 189 | 64 | |
GenBank mRNA | BC134475.1 | - | 1 - 2 | 190 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |