Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTF2IRD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083745.1 | NP_001077214.1 | 4 - 5 | 541 | 95 | |
GenBank mRNA | BC134477.1 | - | 4 - 5 | 541 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |