Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083759.1 | NP_001077228.1 | 3 - 4 | 523 | 72 | |
GenBank mRNA | BC134684.1 | - | 3 - 4 | 523 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |