Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SECTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083787.1 | NP_001077256.1 | 4 - 5 | 553 | 82 | |
GenBank mRNA | BC134744.1 | - | 4 - 5 | 553 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |