Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STX1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083798.1 | NP_001077267.1 | 5 - 6 | 466 | 103 | |
GenBank mRNA | BC123656.1 | - | 5 - 6 | 466 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |