Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXCL17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083799.2 | NP_001077268.1 | 1 - 2 | 304 | 83 | |
GenBank mRNA | BC134719.1 | - | 1 - 2 | 306 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |