Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097561.1 | NP_001091030.1 | 3 - 4 | 237 | 109 | |
GenBank mRNA | BC140489.1 | - | 3 - 4 | 237 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |