Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097572.1 | NP_001091041.1 | 1 - 2 | 127 | 122 | |
GenBank mRNA | BC140658.1 | - | 1 - 2 | 127 | 122 | |
BT030694.1 | - | 1 - 2 | 165 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |