Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100293.2 | NP_001093763.1 | 1 - 2 | 43 | 71 | |
GenBank mRNA | BC147895.1 | - | 1 - 2 | 44 | 71 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Bos taurus Inventoried Probe spans exons |