Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFAF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101049.2 | NP_001094519.2 | 6 - 7 | 740 | 113 | |
GenBank mRNA | BC149742.1 | - | 2 - 3 | 183 | 113 | |
EE333289.1 | - | 6 - 7 | 698 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |