Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFAF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101049.2 | NP_001094519.2 | 7 - 8 | 853 | 89 | |
GenBank mRNA | BC149742.1 | - | 3 - 4 | 296 | 89 | |
EE333289.1 | - | 7 - 8 | 811 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |