Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST8SIA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103227.1 | NP_001096697.1 | 2 - 3 | 582 | 89 | |
GenBank mRNA | AJ868431.1 | - | 2 - 3 | 199 | 89 | |
BC140641.1 | - | 2 - 3 | 582 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |