Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST8SIA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103227.1 | NP_001096697.1 | 4 - 5 | 762 | 109 | |
GenBank mRNA | AJ868431.1 | - | 4 - 5 | 379 | 109 | |
BC140641.1 | - | 4 - 5 | 762 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |