Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113240.1 | NP_001106711.1 | 6 - 7 | 940 | 111 | |
NM_174113.2 | NP_776538.1 | 6 - 7 | 940 | 111 | ||
GenBank mRNA | BC142250.1 | - | 6 - 7 | 1012 | 111 | |
X51689.1 | - | 915 | 111 | |||
X54183.1 | - | 929 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |