Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COQ7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113241.1 | NP_001106712.1 | 1 - 2 | 132 | 95 | |
GenBank mRNA | BC109578.1 | - | 1 - 2 | 96 | 95 | |
DT837410.1 | - | 1 - 2 | 133 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |