Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPPP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173976.3 | NP_776401.1 | 2 - 3 | 464 | 87 | |
GenBank mRNA | BC133343.1 | - | 3 - 4 | 818 | 87 | |
X85738.1 | - | 541 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |