Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGLYRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174573.3 | NP_776998.1 | 1 - 2 | 297 | 93 | |
GenBank mRNA | AY083309.1 | - | 1 - 2 | 299 | 93 | |
BC142042.1 | - | 1 - 2 | 386 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |