Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACVR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176663.3 | NP_788836.2 | 6 - 7 | 966 | 111 | |
GenBank mRNA | BC133311.1 | - | 6 - 7 | 966 | 111 | |
U58095.1 | - | 6 - 7 | 921 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |