Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DBIL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178322.3 | NP_847892.1 | 1 - 2 | 172 | 64 | |
GenBank mRNA | BC103431.1 | - | 1 - 2 | 148 | 64 | |
DT858438.1 | - | 1 - 2 | 173 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |