Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024533.2 | NP_001019704.2 | 7 - 8 | 797 | 77 | |
GenBank mRNA | BC119980.1 | - | 7 - 8 | 797 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |