Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JMJD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034320.2 | NP_001029492.2 | 3 - 4 | 960 | 89 | |
GenBank mRNA | BC104591.1 | - | 3 - 4 | 895 | 89 | |
BT021521.1 | - | 3 - 4 | 960 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |