Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035029.2 | NP_001030201.2 | 3 - 4 | 460 | 92 | |
GenBank mRNA | BC110017.1 | - | 3 - 4 | 460 | 92 | |
BT021548.1 | - | 3 - 4 | 444 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |